Please login to your online account to display your discounted pricing
BioCision™ Hinged Cryoboxes
Standardizes cryogenic sample storage
$74.16 - $629.26
Specifications
Product Type Cryogenic Storage Box
Height (English) 2 in.
Height (Metric) 5cm
Length (English) 5.25 in.
Length (Metric) 13.3cm
View More Specs
Products
Catalog Number Mfr. No. Color Quantity Price Quantity    
13-901-101
biocision™
BCS206B
Blue 5/Pk. Pack of 5 Each for $74.16
13-901-100
biocision™
BCS206
White 5/Pk. Pack of 5 Each for $74.16
13-901-103
biocision™
BCS206O
Orange 5/Pk. Pack of 5 Each for $74.16
13-901-102
biocision™
BCS206G
Green 5/Pk. Pack of 5 Each for $74.16
13-901-105
biocision™
BCS206PK
Pink 5/Pk. Pack of 5 Each for $74.16
13-901-106
biocision™
BCS207
White 50/Pk. Pack of 50 Each for $629.26
13-901-108
biocision™
BCS207G
Green 50/Pk. Pack of 50 Each for $629.26
13-901-107
biocision™
BCS207B
Blue 50/Pk. Pack of 50 Each for $629.26
13-901-109
biocision™
BCS207O
Orange 50/Pk. Pack of 50 Each for $629.26
13-901-104
biocision™
BCS206P
Purple 5/Pk. Pack of 5 Each for $74.16
13-901-110
biocision™
BCS207P
Purple 50/Pk. Pack of 50 Each for $629.26
13-901-111
biocision™
BCS207PK
Pink 50/Pk. Pack of 50 Each for $629.26
13-901-115
biocision™
BCS206MC
Blue, Green, Orange, Pink, Purple 5/Pk. Pack of 5 Each for $74.38
Description & Specifications
Specifications
Product Type Cryogenic Storage Box
Height (English) 2 in.
Height (Metric) 5cm
Length (English) 5.25 in.
Length (Metric) 13.3cm
Material Laminated Cardboard
Width (English) 5.25 in.
Width (Metric) 13.3cm
Includes Hinged lid, 81-place adjustable plastic grid
  • Dual indexing systems
  • Easy open lid minimizes contamination
  • Laminated, waterproof fiberboard
  • Adjustable, removable slats
  • Compatible with 1mL and 2 mL cryovials, microfuge tubes
  • Designed for repeated freezing and thawing
Dimensions: 5.25 x 5.25 x 2 in. (W x L x H)