Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ SEC 120 LC Columns


Outstanding size separation of water soluble compounds.

Manufacturer: thermo scientific™  73405157846

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-051-829

  • / Each
 

Description & Specifications

Specifications

Column Format High Temperature Column
Diameter (Metric) 7.8mm
Pore Size 120Å
pH -
Packing Material Spherical, fully porous ultrapure silica
Temperature -
Surface Area -
USP Type L33
Particle Size 5µm
Stationary Phase Diol
Column Type Size Exclusion
Carbon Load -
For Use With Size Exclusion Chromatography
Length (Metric) 150mm
Endcapped -
Type High Temperature Column
Max. Pressure 400bar

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 LC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size

Ordering Information:

For information on bulk quantities, please inquire.