Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ SEC 1000 LC Columns


Offer outstanding size separation of water soluble compounds with BioBasic™ SEC 1000 LC Columns.

Manufacturer: thermo scientific™  73605157846

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-051-839

  • / Each
 

Description & Specifications

Specifications

Column Format High Temperature Column
Diameter (Metric) 7.8mm
Column Type Normal-Phase
Max. Pressure 400bar
Particle Size 5µm
Temperature -
Pore Size 1000Å
For Use With LC/MS
Stationary Phase BioBasic SEC
Surface Area -
Type High Temperature Column
Length (Metric) 150mm
Carbon Load -
USP Type L33
Packing Material Spherical, fully porous ultrapure silica
pH -
Endcapped -

Offer outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 LC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weight from 100 to 10,000,000 daltons allowing the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Ideal for sample cleanup
  • Straightforward method development, simple mobile phases
  • 1000Å pore size

Ordering Information:

For information on bulk quantities, please inquire.