Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ SCX LC Columns


BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.

Manufacturer: thermo scientific™  73205053030

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-051-801

  • / Each
 

Description & Specifications

Specifications

Column Format Analytical Column
Diameter (Metric) 3mm
Column Type Cation Exchange
Max. Pressure 400bar
Packing Material Spherical, fully porous ultrapure silica
Particle Size 5µm
Temperature 60°C
Pore Size 300Å
pH 2 - 8
For Use With LC/MS, APCI, ESI
Stationary Phase BioBasic SCX
Surface Area 100m²/g
Type Analytical Column
Length (Metric) 50mm
Carbon Load 3%
USP Type L52
Endcapped No

Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.

  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.