Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ 8 LC Columns


Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

Manufacturer: thermo scientific™  72205052130

 View more versions of this product

Catalog No. 03-051-677

  • / Each
 

Description & Specifications

Specifications

Column Type Reversed Phase
Type Analytical Column
Carbon Load 5%
USP Type L7
Endcapped Yes
Max. Pressure 400bar
Packing Material Spherical, fully porous ultrapure silica
Particle Size 5µm
Temperature 60°C
Pore Size 300Å
pH 2 to 9
Stationary Phase C8
Surface Area 100m²/g
Length (Metric) 50mm
Column Format Analytical Column
Diameter (Metric) 2.1mm

Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call Customer Service for more information.