Please login to your online account to display your discounted pricing

Thermo Scientific™ BioBasic™ 4 LC Columns


BioBasic 4 LC Columns are designed for the analysis of larger proteins and peptides.

Manufacturer: thermo scientific™  72305102130

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-051-710

  • / Each
 

Description & Specifications

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1mm
Column Type Reversed Phase
Max. Pressure 400bar
Packing Material Spherical, fully porous ultrapure silica
Particle Size 5µm
Temperature 60°C
Pore Size 300Å
pH 2 - 10
For Use With LC/MS
Stationary Phase BioBasic 4
Surface Area 100m²/g
Type Analytical Column
Length (Metric) 100mm
Carbon Load 4%
USP Type L26
Endcapped Yes

Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

  • Based on 300Å silica for outstanding biomolecule performance
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.