Bio-Serv™ Pregnant/Weanling, High Protein Lieber-DeCarli '82 Rodent Liquid Diet
For convenient measurement of food intake or study of nutritional manipulation
Manufacturer:  Bio-Serv™ F1264
 View more versions of this product
Catalog No. 14726635
Requires : Blender required for mixing.
Description
Description
  • Liquid format allows for the addition of alcohol, medications, or test compounds
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Specifications
Specifications
Liquid
Rodent
1000
Liquid
4L
Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Rodent liquid diet- Control
Ideal for post operative recovery
Documents