Please login to your online account to display your discounted pricing

Bio-Serv™ Pregnant/Weanling, High Protein Lieber-DeCarli '82 Rodent Liquid Diet

For convenient measurement of food intake or study of nutritional manipulation

$78.37 - $78.37

Specifications

Calories 1000
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Quantity 4L
Format Liquid
For Use With (Application) Ideal for post operative recovery
View More Specs
Requires:  Blender required for mixing.
Products
Catalog Number Mfr. No. Product Type Price Quantity    

14726636

 
bio-serv
F1265
Rodent liquid diet- Ethanol Each for $78.37

14726635

 
bio-serv
F1264
Rodent liquid diet- Control Each for $78.37
Description & Specifications

Specifications

Calories 1000
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Quantity 4L
Format Liquid
For Use With (Application) Ideal for post operative recovery
Particle Shape Liquid
Shelf Life 6 Months
Species Rodent

  • Liquid format allows for the addition of alcohol, medications, or test compounds
  • Ideal for post-operative recovery
  • Nutritionally assayed

  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

6 Months