Bio-Serv™ Nylabone Rodent Enrichment Device, Certified
Provides gnawing opportunity for rodents
$182.35 - $282.94
Specifications
Autoclavable Yes
Product Type Nylabones
For Use With (Application) Helps reduce malocclusions and prevent stereotypical behavior
Material Nylon
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Flavor Length (English) Size Width (English) Price Quantity  
Catalog Number Mfr. No. Flavor Length (English) Size Width (English) Price Quantity  
14726507 Bio-Serv
K3200
Natural 3.75 in. Petite 1 in. Each for $182.35
14726559 Bio-Serv
K3201**
Natural 4.5 in. Regular 1.25 in. Pack of 50 Each for $282.94
Specifications
Description
  • Helps reduce malocclusions
  • Helps prevent stereotypical behavior
  • Excellent for rats on powdered chow or liquid diets
  • Made of solid nylon
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Yes
Nylabones
Helps reduce malocclusions and prevent stereotypical behavior
Nylon