missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bio-Serv™ Mouse Diet, High Fat, Soft Pellets

For induction and/or maintenance of obese models

Manufacturer:  Bio-Serv™ F3282

 View more versions of this product

Catalog No. 14726510



Description

Description

  • Nutritionally complete purified diet formula successfully used in diabetes and obesity studies
  • 60% kcals from fat
  • Soft pellets are 0.5 in. dia. (1.3cm) and vary in length
  • Nutritionally assayed

  • Ingredients: Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride

Specifications

Specifications

Soft Pellet
5000g
Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
0.5 in.
1.27cm
Nutritionally complete purified diet formula successfully used in diabetes and obesity studies
Non-sterile
Mouse
5.49
Mouse diet
Pellet
Documents