missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bio-Serv™ Liquid Diet Feeding Tubes

Designed specifically for liquid diets—eliminates clogging that occurs with sipper tubes

Manufacturer:  Bio-Serv™ 9019*

 View more versions of this product

Catalog No. 14-726-518



Requires : For use with stainless steel tube holders (Mfr. Nos. 9015 and 9016)

Description

Description

  • Made of borosilicate glass with autoclavable screw cap
  • Easy-to-read calibrations for accurate food intake
  • Suitable for use with most cages
  • Autoclavable
  • Cagewasher safe

Specifications

Specifications

Yes
0.05L
0.95cm
1.25 in.
For liquid diet feeding
0.013 gal.
3.75 in.
Borosilicate Glass
0.318cm
Liquid diet feeding tube
Documents