Bio-Serv™ Kahn-Pregnant/Weanling Rodent Liquid Diet
Versatile liquid format allows the addition of alcohol, medications, or test compounds
Manufacturer:  Bio-Serv™ F0503
 View more versions of this product
Catalog No. 14726627
Requires : Blender required for mixing.
Description
Description
  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Specifications
Specifications
Powder
Rodent
1000
Powder
4L
Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Rodent liquid diet- Control
Ideal for post operative recovery
Documents