Please login to your online account to display your discounted pricing

Bio-Serv™ Kahn-Pregnant/Weanling Rodent Liquid Diet

Versatile liquid format allows the addition of alcohol, medications, or test compounds

$109.69 - $109.69

Specifications

Calories 1000
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Quantity 4L
Format Powder
For Use With (Application) Ideal for post operative recovery
View More Specs
Requires:  Blender required for mixing.
Products
Catalog Number Mfr. No. Product Type Price Quantity    

14726628

 
bio-serv
F0504
Rodent liquid diet- Ethanol Each for $109.69

14726627

 
bio-serv
F0503
Rodent liquid diet- Control Each for $109.69
Description & Specifications

Specifications

Calories 1000
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Quantity 4L
Format Powder
For Use With (Application) Ideal for post operative recovery
Particle Shape Powder
Shelf Life 6 Months
Species Rodent

  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder

  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

6 Months