Bio-Serv™ Holder for Liquid Diet Feeding Tubes
Adjustable clip allows for variations in tube height
Manufacturer:  Bio-Serv™ 9015**
 View more versions of this product
Catalog No. 14-726-519
Description
Description
  • Stainless steel
  • Fits over lip of cage
  • Autoclavable
  • Cagewasher safe
Specifications
Specifications
Yes
1.21cm
1. 62 in.
For liquid diet feeding
4.75 in.
Stainless Steel
0.254cm
Tube holder
Documents