Bio-Serv™ Enrichment Device, Wood Gnawing Blocks, Certified
Made of high quality, kiln-dried hardwood
Manufacturer:  Bio-Serv™ K3511
 View more versions of this product
Catalog No. 14726506
Description
Description
  • 100% natural wood—show variations in wood grain patterns and color
  • Provides a natural form of gnawing enrichment
  • Cube shape reduces loss through cage floor
  • Autoclavable
  • Certified (Contaminant screened)
Specifications
Specifications
Yes
Provide the most natural form of gnawing enrichment
3.17cm
3.17cm
Small
3.17cm
1.25 x 1.25 x 1.25 in. (3.17 x 3.17 x 3.17cm)
1.25 in.
1.25 in.
Kiln Dried Hardwood
1.25 in.
Wood gnawing block
Documents