Please login to your online account to display your discounted pricing
Bio-Serv™ Enrichment Device, Wood Gnawing Blocks, Certified
Made of high quality, kiln-dried hardwood
$166.73 - $200.88
Specifications
Autoclavable Yes
Product Type Wood gnawing block
For Use With (Application) Provide the most natural form of gnawing enrichment
Material Kiln Dried Hardwood
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
14726575
bio-serv
K3512
Medium Pack of 112 Each for $166.73
14726576
bio-serv
K3513
Large Pack of 54 Each for $200.88
Description & Specifications
Specifications
Autoclavable Yes
Product Type Wood gnawing block
For Use With (Application) Provide the most natural form of gnawing enrichment
Material Kiln Dried Hardwood
  • 100% natural wood—show variations in wood grain patterns and color
  • Provides a natural form of gnawing enrichment
  • Cube shape reduces loss through cage floor
  • Autoclavable
  • Certified (Contaminant screened)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.