Bio-Serv™ Enrichment Device, Wood Gnawing Blocks, Certified
Made of high quality, kiln-dried hardwood
Manufacturer:  Bio-Serv™ K3512
 View more versions of this product
Catalog No. 14726575
Description
Description
  • 100% natural wood—show variations in wood grain patterns and color
  • Provides a natural form of gnawing enrichment
  • Cube shape reduces loss through cage floor
  • Autoclavable
  • Certified (Contaminant screened)
Specifications
Specifications
Yes
Provide the most natural form of gnawing enrichment
3.81cm
3.81cm
Medium
3.81cm
1.5 x 1.5 x 1.5 in. (3.81 x 3.81 x 3.81cm)
1.5 in.
1.5 in.
Kiln Dried Hardwood
1.5 in.
Wood gnawing block
Documents