Bio-Serv™ AIN-76 Rodent Liquid Animal Diet
No blender needed—just add water, shake, and pour
Manufacturer:  Bio-Serv™ F1268
 View more versions of this product
Catalog No. 14726625
Description
Description
  • Easy to prepare
  • Offers the versatility of a liquid format, allowing the addition of alcohol, medications, or test compounds
  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Corn Oil, Cellulose, Maltodextrin, Mineral Mix, Suspending Aids, Vitamin Mix, DL-Methionine, Choline Bitartrate
Specifications
Specifications
Pellet
Rodent
Rodent Liquid Diet, Control
Post operative recovery
4L
Corn syrup solids, Casein, Maltodextrin, Corn Oil, Cellulose, Mineral Mix, Suspending Aid, Vitamin Mix, DL-Methionine, Choline Bitartrate
Powder
Documents