Please login to your online account to display your discounted pricing

BIO-acetoxime, Tocris™

Selective GSK-3α/β inhibitor

$168.15 - $362.13

Chemical Identifiers

Chemical Name or Material BIO-acetoxime
Assay Percent Range >98%
Molecular Formula C18H12BrN3O3
Formula Weight 398.21
Synonym (2'Z, 3'E)-6-Bromoindirubin-3'-acetoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

38741

 
r & d systems
RDS38741
1mg Each for $168.15

387410

 
r & d systems
RDS387410
10mg Each for $362.13
Description & Specifications

Specifications

Formula Weight 398.21
Quantity 1mg
Chemical Name or Material BIO-acetoxime
Assay Percent Range >98%
Molecular Formula C18H12BrN3O3
CAS 667463-85-6
Synonym (2'Z, 3'E)-6-Bromoindirubin-3'-acetoxime
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C