Whatman™ Binder-Free Glass Microfiber Filters: PROMO OFFER
Offers fine particle retention
$100.30 - $100.30
Specifications
Material Glass Microfiber
Grade GF/A
Particle Retention 1.6μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Format Product Type Price Quantity  
Catalog Number Mfr. No. Format Product Type Price Quantity  
0987416CPM Whatman™
1820110PM
Circle Circle; Diameter: 11.0cm Pack of 100 Each for $100.30
Specifications
Description
  • High flowrate
  • Good loading capacity
  • Water pollution effluent monitoring
  • Filtering water, algae, bacterial cultures
  • Foodstuff analyses
  • Protein filtration
  • Radioimmunoassay of weak β-emitters
  • Gravimetric determination of airborne particulates, stack sampling and absorption methods of air pollution monitoring
Description & Specifications
Specifications
Glass Microfiber
GF/A
1.6μm