missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BIMU 8, Tocris™

Potent 5-HT4 receptor full agonist

$232.08 - $956.46

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C19H26N4O2·HCl
Formula Weight 378.9
Synonym 2, 3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride
Chemical Name or Material BIMU 8
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
437410 R & D Systems
437410
10mg Each for $232.08
437450 R & D Systems
437450
50mg Each for $956.46
Specifications Description & Specifications

Chemical Identifiers

>98%
378.9
BIMU 8
C19H26N4O2·HCl
2, 3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride

Specifications

10mg
>98%
378.9
BIMU 8
134296-40-5
C19H26N4O2·HCl
2, 3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride

Safety and Handling

Recommended Storage : Store at +4°C