Please login to your online account to display your discounted pricing

BIIE 0246, Tocris

Potent, selective non-peptide NPY Y2 receptor antagonist

$189.01 - $401.24

Chemical Identifiers

Chemical Name or Material BIIE 0246
Assay Percent Range >98%
Molecular Formula C49H57N11O6
Formula Weight 896.06
Synonym N-[(1S)-4-[(Aminoiminomethyl)amino]-1-[[[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]amino]carbonyl]butyl]-1-[2-[4-(6,11-dihydro-6-oxo-5H-dibenz[b,e]azepin-11-yl)-1-piperazinyl]-2-oxoethyl]-cyclopentaneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

17001

 
r & d systems
17001
1mg Each for $189.01

170010

 
r & d systems
170010
10mg Each for $401.24
Description & Specifications

Specifications

Formula Weight 896.06
Quantity 1mg
CAS 246146-55-4
Chemical Name or Material BIIE 0246
Assay Percent Range >98%
Molecular Formula C49H57N11O6
Synonym N-[(1S)-4-[(Aminoiminomethyl)amino]-1-[[[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]amino]carbonyl]butyl]-1-[2-[4-(6,11-dihydro-6-oxo-5H-dibenz[b,e]azepin-11-yl)-1-piperazinyl]-2-oxoethyl]-cyclopentaneacetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C