Please login to your online account to display your discounted pricing

BIIB 021, Tocris™

Selective Hsp90 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material BIIB 021
Assay Percent Range >98%
Molecular Formula C14H15ClN6O
Formula Weight 318.76
Synonym 6-Chloro-9-[(4-methoxy-3, 5-dimethyl-2-pyridinyl)methyl]-9H-purin-2-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

460810

 
r & d systems
460810
10mg Each for $252.39

460850

 
r & d systems
460850
50mg Each for $1,050.42
Description & Specifications

Specifications

Chemical Name or Material BIIB 021
Quantity 10mg
CAS 848695-25-0
Recommended Storage Store at -20°C
Assay Percent Range >98%
Molecular Formula C14H15ClN6O
Formula Weight 318.76
Synonym 6-Chloro-9-[(4-methoxy-3, 5-dimethyl-2-pyridinyl)methyl]-9H-purin-2-amine
Safety and Handling

Store at -20°C