Please login to your online account to display your discounted pricing

Alfa Aesar™ Big Endothelin-2 (22-38), Human

For cellular and molecular biology applications

Specifications

Product Type Big Endothelin-2 (22-38), human
Molecular Weight 1810.96
Storage Requirements -30° to -10°C
Sequence Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66163EXD

 
alfa aesar™
J66163EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66163MCR

 
alfa aesar™
J66163MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Big Endothelin-2 (22-38), human
Molecular Weight 1810.96
Storage Requirements -30° to -10°C
Sequence Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC