Please login to your online account to display your discounted pricing

Alfa Aesar™ Big Endothelin-2 (22-37), Human

For cellular and molecular biology applications

Specifications

Product Type Big Endothelin-2 (22-37), human
Cross Reactivity Human
Molecular Weight 1654.77
Storage Requirements -30° to -10°C
Sequence Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66002EXD

 
alfa aesar™
J66002EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66002MCR

 
alfa aesar™
J66002MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Big Endothelin-2 (22-37), human
Cross Reactivity Human
Molecular Weight 1654.77
Storage Requirements -30° to -10°C
Sequence Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro
Format Solid
Regulatory Status RUO
Purification Method HPLC