Please login to your online account to display your discounted pricing

Alfa Aesar™ Big Endothelin-1 (22-38), Human

For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66781-MCR

 View all options for this product

Catalog No. AAJ66781MCR

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Molecular Weight 1808.98
Purification Method HPLC
Quantity 1mg
Format Solid
Sequence Val-Asn-Thr-Pro-Glu-His-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Arg-Ser
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Big Endothelin-1 (22-38), human