Learn More
Bicyclo[3.2.0]hept-2-en-6-one, 97%, Thermo Scientific Chemicals
$202.92 - $2058.90
Chemical Identifiers
CAS | 13173-09-6 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00066201 |
InChI Key | LNLLHUHPGPKRBM-UHFFFAOYSA-N |
Synonym | bicyclo 3.2.0 hept-2-en-6-one, +/--cis-bicyclo 3.2.0 hept-2-en-6-one, cis-bicyclo 3.2.0 hept-2-en-6-one, 1s,5s-bicyclo 3.2.0 hept-2-en-6-one, +-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-ene-6-one, bicyclo 3.2.0 hept-3-en-7-one, z-bicyclo 3.2.0 hepta-2-ene-6-one |
PubChem CID | 297183 |
IUPAC Name | bicyclo[3.2.0]hept-3-en-7-one |
SMILES | C1C=CC2C1C(=O)C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2247206
|
Thermo Scientific Chemicals
B2247206 |
5 g |
Each for $202.92
|
|
|||||
AAB2247222
|
Thermo Scientific Chemicals
B2247222 |
100 g |
Each for $2,058.90
|
|
|||||
Description
Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
13173-09-6 | |
108.14 | |
LNLLHUHPGPKRBM-UHFFFAOYSA-N | |
297183 | |
C1C=CC2C1C(=O)C2 |
C7H8O | |
MFCD00066201 | |
bicyclo 3.2.0 hept-2-en-6-one, +/--cis-bicyclo 3.2.0 hept-2-en-6-one, cis-bicyclo 3.2.0 hept-2-en-6-one, 1s,5s-bicyclo 3.2.0 hept-2-en-6-one, +-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-ene-6-one, bicyclo 3.2.0 hept-3-en-7-one, z-bicyclo 3.2.0 hepta-2-ene-6-one | |
bicyclo[3.2.0]hept-3-en-7-one |
Specifications
13173-09-6 | |
MFCD00066201 | |
UN1224 | |
bicyclo 3.2.0 hept-2-en-6-one, +/--cis-bicyclo 3.2.0 hept-2-en-6-one, cis-bicyclo 3.2.0 hept-2-en-6-one, 1s,5s-bicyclo 3.2.0 hept-2-en-6-one, +-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-ene-6-one, bicyclo 3.2.0 hept-3-en-7-one, z-bicyclo 3.2.0 hepta-2-ene-6-one | |
C1C=CC2C1C(=O)C2 | |
108.14 | |
108.14 | |
Bicyclo[3.2.0]hept-2-en-6-one |
C7H8O | |
5 g | |
Air Sensitive | |
LNLLHUHPGPKRBM-UHFFFAOYSA-N | |
bicyclo[3.2.0]hept-3-en-7-one | |
297183 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only