Please login to your online account to display your discounted pricing

Alfa Aesar™ Bicyclo[2.2.2]oct-2-ene-2,3-dicarboxylic anhydride, 98%

$76.39 - $308.89

Chemical Identifiers

Chemical Name or Material Bicyclo[2.2.2]oct-2-ene-2,3-dicarboxylic anhydride
CAS 151813-29-5
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD01318101
Beilstein 167495
Synonym 4,5,6,7-Tetrahydro-4,7-ethanoisobenzofuran-1,3-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1534803

  SDS
alfa aesar™
L1534803
1g Each for $76.39

AAL1534806

  SDS
alfa aesar™
L1534806
5g Each for $308.89
Description & Specifications

Specifications

Formula Weight 178.19
CAS 151813-29-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H10O3
MDL Number MFCD01318101
Chemical Name or Material Bicyclo[2.2.2]oct-2-ene-2,3-dicarboxylic anhydride
Synonym 4,5,6,7-Tetrahydro-4,7-ethanoisobenzofuran-1,3-dione
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Beilstein 167495
Melting Point 158° to 160°C
Assay 98%

Applications
Bicyclo[2.2.2]oct-2-ene-2,3-dicarboxylic anhydride is used as primary and secondary intermediate in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.