Rotating Platform
Manufacturer:  Fisher Science Education EPD40226
Catalog No. S94222
Description
Description
Use this wide, stable, rotating platform and bicycle wheel gyroscope (avaible separately) to turn into a human gyroscope.
Platform features:
  • Wood and steel construction
  • Sturdy with nonslip pads for traction
  • Can be used to demonstrate rolling friction, precession of a gyroscope, weather demonstrations and rotational inertia
  • Diameter: 16 in. (40cm)
Order Info
Bicycle wheel gyroscope is not included
Specifications
Specifications
16 in.
40cm
Can be used to demonstrate rolling friction, precession of a gyroscope, weather demonstrations and rotational inertia
Rotating platform