Please login to your online account to display your discounted pricing
Bicycle Wheel Gyroscope and Rotating Platform
Bring gyroscopic forces to life.
$115.05 - $115.05
Specifications
For Use With (Application) Can be used to demonstrate rolling friction, precession of a gyroscope, weather demonstrations and rotational inertia
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S94222
fisher science education
40-226
Rotating platform Each for $115.05
Description & Specifications
Specifications
For Use With (Application) Can be used to demonstrate rolling friction, precession of a gyroscope, weather demonstrations and rotational inertia
Use this wide, stable, rotating platform and bicycle wheel gyroscope together to turn into a human gyroscope.
Wheel features:
  • Turns freely on axis
  • Equipped with large, comfortable handles at each end
  • Mass concentrated at rim makes rotation more apparent as wheel tilts
  • Includes instructions with experiments
Platform features:
  • Wood and steel construction
  • Sturdy with nonslip pads for traction
  • Can be used to demonstrate rolling friction, precession of a gyroscope, weather demonstrations and rotational inertia
  • Diameter: 16 in. (40cm)