Please login to your online account to display your discounted pricing

(-)-Bicuculline methochloride, Tocris


Water-soluble GABAA antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS013110

 View more versions of this product

Catalog No. 013110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 417.85
CAS 53552-05-9
Chemical Name or Material (-)-Bicuculline methochloride
Assay Percent Range >99%
Molecular Formula C21H20ClNO6
Synonym [R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium chloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (-)-Bicuculline methochloride
Assay Percent Range >99%
Molecular Formula C21H20ClNO6
Formula Weight 417.85
Synonym [R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium chloride
Accessories

Store at Room Temperature