missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Bicuculline methochloride, Tocris

Water-soluble GABAA antagonist

$149.90 - $581.39

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C21H20ClNO6
Formula Weight 417.85
Synonym [R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium chloride
Chemical Name or Material (-)-Bicuculline methochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
013110 R & D Systems
RDS013110
10mg Each for $149.90
013150 R & D Systems
RDS013150
50mg Each for $581.39
Specifications Description & Specifications

Chemical Identifiers

>99%
417.85
(-)-Bicuculline methochloride
C21H20ClNO6
[R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium chloride

Specifications

53552-05-9
C21H20ClNO6
10mg
(-)-Bicuculline methochloride
>99%
417.85
[R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium chloride

Safety and Handling

Recommended Storage : Store at Room Temperature