Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-Bicuculline methiodide, 98%

Chemical Identifiers

Chemical Name or Material (-)-Bicuculline methiodide
CAS 40709-69-1
Formula Weight 509.3
MDL Number MFCD00078966
UN Number UN1544
Beilstein 5419649
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67150MF

  SDS
alfa aesar™
J67150MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 509.3
Chemical Name or Material (-)-Bicuculline methiodide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H330
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Physical Form Solid
Assay 98%
MDL Number MFCD00078966
Color Pale yellow
Beilstein 5419649
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
CAS 40709-69-1
UN Number UN1544
Quantity 50mg
RTECS Number JI4904100
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H330
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.