Please login to your online account to display your discounted pricing

Bicine, Fisher BioReagents


Quantity 250g
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP2646250

 View more versions of this product

Catalog No. BP2646250

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 190°C
Physical Form Powder/Solid
Formula Weight 163.2
Quantity 250g
Merck Index 15, 1205
pH 4.0 to 5.0
Color White
Molecular Formula C6H13NO4
Chemical Name or Material Bicine
Absorbance 0.100 AU max. at 260nm
Packaging Poly Bottle
Linear Formula (HOCH2CH2)2NCH2COOH
CAS 150-25-4
ChemAlert Storage Symbol Gray
Synonym N, N-bis [2-hydroxyethyl]glycine
FTIR Conforms to reference
Recommended Storage RT

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Physical Form Powder/Solid
Molecular Formula C6H13NO4
Linear Formula (HOCH2CH2)2NCH2COOH
Formula Weight 163.2
Synonym N, N-bis [2-hydroxyethyl]glycine
Accessories
 

RT

SDS & Safety Documents