Please login to your online account to display your discounted pricing

Bicine, Fisher BioReagents


Quantity 100g
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP2646100

 View more versions of this product

Catalog No. BP2646100

  • / Each
  • / Case of 3 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Melting Point 190°C
Formula Weight 163.2
Quantity 100g
Merck Index 15, 1205
Molecular Formula C6H13NO4
pH 4.0 to 5.0
Color White
Absorbance 0.100 AU max. at 260nm
CAS 150-25-4
Chemical Name or Material Bicine
Recommended Storage RT
Packaging Poly Bottle
FTIR Conforms to reference
Synonym N, N-bis [2-hydroxyethyl]glycine
Linear Formula (HOCH2CH2)2NCH2COOH
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Physical Form Powder/Solid
Molecular Formula C6H13NO4
Linear Formula (HOCH2CH2)2NCH2COOH
Formula Weight 163.2
Synonym N, N-bis [2-hydroxyethyl]glycine
Accessories
 

RT

SDS & Safety Documents