Please login to your online account to display your discounted pricing

Bicine, Fisher BioReagents


Quantity 1kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP26461

 View more versions of this product

Catalog No. BP26461

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 190°C
Physical Form Powder/Solid
Formula Weight 163.2
Quantity 1kg
pH 4.0 to 5.0
Color White
Molecular Formula C6H13NO4
Merck Index 15, 1205
Linear Formula (HOCH2CH2)2NCH2COOH
CAS 150-25-4
ChemAlert Storage Symbol Gray
Recommended Storage RT
Absorbance 0.100 AU max. at 260nm
Chemical Name or Material Bicine
Synonym N, N-bis [2-hydroxyethyl]glycine
FTIR Conforms to reference
Packaging Poly Bottle

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Physical Form Powder/Solid
Molecular Formula C6H13NO4
Linear Formula (HOCH2CH2)2NCH2COOH
Formula Weight 163.2
Synonym N, N-bis [2-hydroxyethyl]glycine
Accessories
 

RT

SDS & Safety Documents