missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bicine, Fisher BioReagents

$586.30 - $586.30

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP2646250
SDS View Documents Promotion Details
Fisher BioReagents
BP2646250
250 g Poly Bottle
Each for $586.30
Case of 4 Each for $2,345.20
 
Description

Description

Bicine is one of the zwitterionic amino acids known as Good buffers. Used as a biological buffer and chelating agent.
Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
8761
2-[bis(2-hydroxyethyl)amino]acetic acid
C6H12NNaO4
MFCD00004295
N, N-bis [2-hydroxyethyl]glycine
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O

Specifications

0.100 AU max. at 260nm, 0.100 AU max. at 280nm
150-25-4
4.0 to 5.0
250 g
(HOCH2CH2)2NCH2COOH
Poly Bottle
N, N-bis [2-hydroxyethyl]glycine
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
Bicine
190°C
White
Powder/Solid
C6H12NNaO4
MFCD00004295
15, 1205
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.2
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
missing translation for 'documents'

missing translation for 'documents'