Please login to your online account to display your discounted pricing

Bicine, 99+%, for biochemistry, ACROS Organics™

$37.32 - $37.32

Chemical Identifiers

Chemical Name or Material Bicine
Name Note For biochemistry
CAS 150-25-4
CAS Min % 99.0
CAS Max % 100.0
Purity Grade For biochemistry
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H13NO4
Linear Formula (HOCH2CH2)2NCH2CO2H
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC327715000

  SDS
thermo fisher scientific
AC327715000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC327711000

  SDS
fisher scientific acros
327711000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC327710250

  SDS
thermo fisher scientific
AC327710250
Plastic bottle 25g Each for $37.32
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 163.17
Assay ≥99.0%
Quantity 500g
Color White
Purity Grade For biochemistry
Iron (Fe) 5ppm max.
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
Name Note For biochemistry
Assay Percent Range 99+%
CAS 150-25-4
Chemical Name or Material Bicine
Molecular Formula C6H13NO4
MDL Number MFCD00004295
CAS Min % 99.0
CAS Max % 100.0
Absorbance 0.05 max. (0.5M, water) at 260nm
Packaging Plastic bottle
Synonym Diethylolglycine
Water 1% max. (K.F.)
Linear Formula (HOCH2CH2)2NCH2CO2H

Biological buffer