Please login to your online account to display your discounted pricing

Bicine, 99+%, for analysis, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC172652500

 View more versions of this product

Catalog No. AC172652500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0% (Total base)
Purity Grade For analysis
Physical Form Crystalline Powder
Formula Weight 163.17
Quantity 250g
Color White
TSCA TSCA
EINECS Number 205-755-1
RTECS Number MB9700000
pH 4.0 to 5.5 (10% soln. at 20°C)
Water 1% Max. (K.F.)
Merck Index 15, 1205
CAS Max % 100.0
CAS Min % 99.0
Sulfated Ash 0.1 % max.
Synonym Diethylolglycine
Linear Formula (HOCH2CH2)2NCH2CO2H
Molecular Formula C6H13NO4
Assay Percent Range 99+%
MDL Number MFCD00004295
Chemical Name or Material Bicine
Name Note 99+%
CAS 150-25-4
Packaging Glass bottle
Lead (Pb) 5ppm max.
Copper (Cu) 2ppm max.
Chlorine (Cl) 50ppm max.
Chloride 50ppm max.
Absorbance Conditions (5% aq. soln.) (1cm cell)
Iron (Fe) 5ppm max.
Absorbance 0.05 max. (5% aqueous soln.) at 250nm
Sulfate 50ppm max.
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Bicine
Name Note 99+%
CAS 150-25-4
CAS Min % 99.0
CAS Max % 100.0
Purity Grade For analysis
Assay ≥99.0% (Total base)
Assay Percent Range 99+%
Molecular Formula C6H13NO4
Linear Formula (HOCH2CH2)2NCH2CO2H
Formula Weight 163.17
MDL Number MFCD00004295
Synonym Diethylolglycine