missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bicine, 99+%, for analysis, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 172650010

Catalog No. AC172650010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Specifications

0.05 max. (5% aqueous soln.) at 250nm
99+%
150-25-4
100.0
1kg
Authentic
(HOCH2CH2)2NCH2CO2H
Glass bottle
N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
≥99.0% (Total base)
Bicine
(5% aq. soln.) (1cm cell)
99.0
4.0 to 5.5 (10% soln. at 20°C)
99+%
C6H12NNaO4
MFCD00004295
15, 1205
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.17
Analysis
Safety and Handling

Safety and Handling

EINECSNumber : 205-755-1

RTECSNumber : MB9700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only