missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ BICINE, 0.5M buffer soln., pH 9.0

$84.96 - $133.08

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H13NO4
Molecular Weight (g/mol) 163.173
MDL Number MFCD00004295
InChI Key FSVCELGFZIQNCK-UHFFFAOYSA-N
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES C(CO)N(CCO)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62838AE View Documents Alfa Aesar™
J62838AE
100mL Each for $84.96
 
AAJ62838AK View Documents Alfa Aesar™
J62838AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

150-25-4
163.173
FSVCELGFZIQNCK-UHFFFAOYSA-N
CHEBI:40957
C(CO)N(CCO)CC(=O)O
C6H13NO4
MFCD00004295
8761
2-[bis(2-hydroxyethyl)amino]acetic acid

Specifications

150-25-4
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
Liquid
100mL
C6H13NO4
0.5M buffer soln.
C(CO)N(CCO)CC(=O)O
163.173
CHEBI:40957
9.0
BICINE buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA