missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ BICINE, 0.5M buffer soln., pH 8.0

$90.22 - $141.73

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H13NO4
Molecular Weight (g/mol) 163.173
MDL Number MFCD00004295
InChI Key FSVCELGFZIQNCK-UHFFFAOYSA-N
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES C(CO)N(CCO)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63924AE View Documents Alfa Aesar™
J63924AE
100mL Each for $90.22
 
AAJ63924AK View Documents Alfa Aesar™
J63924AK
250mL Each for $141.73
 
Description

Description

Applications
BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

150-25-4
163.173
FSVCELGFZIQNCK-UHFFFAOYSA-N
CHEBI:40957
C(CO)N(CCO)CC(=O)O
C6H13NO4
MFCD00004295
8761
2-[bis(2-hydroxyethyl)amino]acetic acid

Specifications

150-25-4
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
Liquid
100mL
C6H13NO4
0.5M buffer soln.
C(CO)N(CCO)CC(=O)O
163.173
CHEBI:40957
8.0
BICINE buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C