Please login to your online account to display your discounted pricing

Alfa Aesar™ Bicine, 0.2M buffer soln., pH 9.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Bicine
CAS 150-25-4
Name Note low endotoxin
MDL Number MFCD00004295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67230AE

 
alfa aesar™
J67230AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67230AK

 
alfa aesar™
J67230AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Quantity 100mL
Name Note low endotoxin
Physical Form Liquid
MDL Number MFCD00004295
pH 9.0
Color Colorless
TSCA TSCA
Concentration 0.2M buffer soln.
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C