Please login to your online account to display your discounted pricing

Alfa Aesar™ Bicine, 0.2M buffer soln., pH 8.5, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67296AK

 View more versions of this product

Catalog No. AAJ67296AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Bicine
MDL Number MFCD00004295
CAS 150-25-4
Physical Form Liquid
TSCA TSCA
Storage Note 1 2° to 8°C
pH 8.5
Color Colorless
Concentration 0.2M buffer soln.
Quantity 250mL
Name Note low endotoxin

Specifications

Chemical Name or Material Bicine
CAS 150-25-4
Name Note low endotoxin
MDL Number MFCD00004295
Accessories

2° to 8°C