missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bicinchoninic acid disodium salt

For determination of Cu and protein assays

Manufacturer:  Alfa Aesar™ 4424709

 View more versions of this product

Catalog No. AA4424709



Specifications

Specifications

Bicinchoninic acid disodium salt
979-88-4
MFCD00037500
Soluble in water.
C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)[O-])C(=O)[O-].[Na+].[Na+]
388.29
388.29
10g
C20H10N2Na2O4
2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt
AUPXFICLXPLHBB-UHFFFAOYSA-L
disodium;2-(4-carboxylatoquinolin-2-yl)quinoline-4-carboxylate
164763
Powder/Chunks

Chemical Identifiers

979-88-4
388.29
AUPXFICLXPLHBB-UHFFFAOYSA-L
164763
C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)[O-])C(=O)[O-].[Na+].[Na+]
C20H10N2Na2O4
MFCD00037500
2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt
disodium;2-(4-carboxylatoquinolin-2-yl)quinoline-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents