Please login to your online account to display your discounted pricing

Bicifadine hydrochloride, Tocris™

Noradrenalin, 5-HT and dopamine re-uptake inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material Bicifadine hydrochloride
Assay Percent Range >98%
Molecular Formula C12H15N·HCl
Formula Weight 209.72
Synonym 1-(4-Methylphenyl)-3-azabicyclo[3.1.0]hexane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

435710

 
r & d systems
435710
10mg Each for $244.81

435750

 
r & d systems
435750
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 209.72
Quantity 10mg
Chemical Name or Material Bicifadine hydrochloride
CAS 66504-75-4
Synonym 1-(4-Methylphenyl)-3-azabicyclo[3.1.0]hexane hydrochloride
Assay Percent Range >98%
Molecular Formula C12H15N·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature