missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bicarbonate, 1M buffer soln., pH 8.0

$84.96 - $109.03

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.006
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES C(=O)(O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63491AK View Documents Alfa Aesar™
J63491AK
250mL Each for $84.96
 
AAJ63491AP View Documents Alfa Aesar™
J63491AP
500mL Each for $109.03
 
Specifications

Chemical Identifiers

144-55-8
84.006
UIIMBOGNXHQVGW-UHFFFAOYSA-M
CHEBI:32139
C(=O)(O)[O-].[Na+]
CHNaO3
MFCD00003528
516892
sodium;hydrogen carbonate

Specifications

144-55-8
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
Liquid
250mL
CHNaO3
1M buffer soln.
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
8.0
Bicarbonate buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA