Please login to your online account to display your discounted pricing

Bicalutamide, Tocris

Non-steroidal androgen receptor antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material Bicalutamide
Assay Percent Range >99%
Molecular Formula C18H14F4N2O4S
Formula Weight 430.37
Synonym N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

338910

 
r & d systems
RDS338910
10mg Each for $244.81

338950

 
r & d systems
RDS338950
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 430.37
Quantity 10mg
Chemical Name or Material Bicalutamide
Assay Percent Range >99%
Molecular Formula C18H14F4N2O4S
CAS 90357-06-5
Synonym N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature