Please login to your online account to display your discounted pricing

BIBX 1382 dihydrochloride, Tocris

Highly selective EGFR-kinase inhibitor

$133.98 - $284.81

Chemical Identifiers

Chemical Name or Material BIBX 1382 dihydrochloride
Assay Percent Range >97%
Molecular Formula C18H19ClFN7·2HCl
Formula Weight 460.77
Synonym N8-(3-Chloro-4-fluorophenyl)-N2-(1-methyl-4-piperidinyl)-pyrimido[5, 4-d]pyrimidine-2, 8-diamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

24161

 
r & d systems
24161
1mg Each for $133.98

241610

 
r & d systems
241610
10mg Each for $284.81
Description & Specifications

Specifications

Formula Weight 460.77
Quantity 1mg
Chemical Name or Material BIBX 1382 dihydrochloride
Assay Percent Range >97%
Molecular Formula C18H19ClFN7·2HCl
CAS 1216920-18-1
Recommended Storage Desiccate at +4°C
Synonym N8-(3-Chloro-4-fluorophenyl)-N2-(1-methyl-4-piperidinyl)-pyrimido[5, 4-d]pyrimidine-2, 8-diamine dihydrochloride
Safety and Handling

Desiccate at +4°C