Please login to your online account to display your discounted pricing

BIBU 1361 dihydrochloride, Tocris

Selective inhibitor of EGFR-kinase

$133.98 - $284.81

Chemical Identifiers

Chemical Name or Material BIBU 1361 dihydrochloride
Assay Percent Range >97%
Molecular Formula C22H27ClFN7·2HCl
Formula Weight 516.87
Synonym N-(3-Chloro-4-fluorophenyl)-6-[4-[(diethylamino)methyl]-1-piperidinyl]-pyrimido[5, 4-d]pyrimidin-4-amine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

241710

 
r & d systems
241710
10mg Each for $284.81

24171

 
r & d systems
24171
1mg Each for $133.98
Description & Specifications

Specifications

Formula Weight 516.87
Quantity 10mg
CAS 793726-84-8
Chemical Name or Material BIBU 1361 dihydrochloride
Synonym N-(3-Chloro-4-fluorophenyl)-6-[4-[(diethylamino)methyl]-1-piperidinyl]-pyrimido[5, 4-d]pyrimidin-4-amine dihydrochloride
Molecular Formula C22H27ClFN7·2HCl
Assay Percent Range >97%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C